Learn More
Electron Microscopy Sciences EMS Wafer Tweezers Style 38FG
SDP
Catalog No. 5024001
Encompass_Preferred
These tweezers are especially designed for the handling of delicate and fragile silicon wafers, scintillation discs, glass slides, coverslips etc. Made from antimagnetic/anti acid steel with a very smooth surface and a non-glare finish.
Tip Width: 9.5 mm
Depth: 11.0 mm
Tip Lip: E:2.5 mm
Catalog No. 50-240-01 Supplier Electron Microscopy Sciences Supplier No. 7841138FG
These tweezers are especially designed for the handling of delicate and fragile silicon wafers, scintillation discs, glass slides, coverslips etc. Made from antimagnetic/anti acid steel with a very smooth surface and a non-glare finish.
Tip Width: 9.5 mm
Depth: 11.0 mm
Tip Lip: E:2.5 mm