Learn More
Electron Microscopy Sciences EMS Wafer Tweezers Style 39S2
SDP
Catalog No. 5024005
Encompass_Preferred
These tweezers are especially designed for the handling of delicate and fragile silicon wafers, scintillation discs, glass slides, coverslips etc. Made from antimagnetic/anti acid steel with a very smooth surface and a non-glare finish.
Tip Width: 6.5 mm
Depth: 8.5 mm
Tip Lip: E:2.4 mm
Offset: 5.5 mm
Catalog No. 50-240-05 Supplier Electron Microscopy Sciences Supplier No. 7841239S2
These tweezers are especially designed for the handling of delicate and fragile silicon wafers, scintillation discs, glass slides, coverslips etc. Made from antimagnetic/anti acid steel with a very smooth surface and a non-glare finish.
Tip Width: 6.5 mm
Depth: 8.5 mm
Tip Lip: E:2.4 mm
Offset: 5.5 mm