Learn More
Electron Microscopy Sciences EMS Tweezers Style 3CB, SA steel, Very sharp, superior finish, bent tips Tips
SDP
Catalog No. 5023852
Encompass_Preferred
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish.
Material: SA Steel
Tips: Very sharp, superior finish, bent tips
OAL: 110 mm
Catalog No. 50-238-52 Supplier Electron Microscopy Sciences Supplier No. 783183CB
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish.
Material: SA Steel
Tips: Very sharp, superior finish, bent tips
OAL: 110 mm