Learn More
Electron Microscopy Sciences EMS Tweezers Style 3CTA, Titanium alloy, Very fine tips and superior finish Tips
SDP
Catalog No. 5023856
Encompass_Preferred
The tweezers are the Swiss highest quality. Titanium alloy with density (4.5g/cm3), good mechanical properties and high temp (1600 deg F/870 deg C) use. Good corrosion resistance. Non-magnetic.
Material: Titanium Alloy
Tips: Very fine tips and superior finish
OAL: 110 mm
Catalog No. 50-238-56 Supplier Electron Microscopy Sciences Supplier No. 783183CTA
The tweezers are the Swiss highest quality. Titanium alloy with density (4.5g/cm3), good mechanical properties and high temp (1600 deg F/870 deg C) use. Good corrosion resistance. Non-magnetic.
Material: Titanium Alloy
Tips: Very fine tips and superior finish
OAL: 110 mm