Learn More
Electron Microscopy Sciences EMS Tweezers Style 3F, SA steel, Parallel touching Tips
SDP
Catalog No. 5023865
Encompass_Preferred
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish. Tips touching over 3mm (parallel tips). Useful for handling tiny wires.
Material: SA Steel
Tips: Parallel touching
OAL: 120 mm
Catalog No. 50-238-65 Supplier Electron Microscopy Sciences Supplier No. 783183F
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish. Tips touching over 3mm (parallel tips). Useful for handling tiny wires.
Material: SA Steel
Tips: Parallel touching
OAL: 120 mm