Learn More
Electron Microscopy Sciences EMS Tweezers Style 3X, SA steel, Very sharp tips, self closing Tips
SDP
Catalog No. 5023860
Encompass_Preferred
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish.
Material: SA Steel
Tips: Very sharp tips, self closing
OAL: 120 mm
Catalog No. 50-238-60 Supplier Electron Microscopy Sciences Supplier No. 783183X
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish.
Material: SA Steel
Tips: Very sharp tips, self closing
OAL: 120 mm