Learn More
Electron Microscopy Sciences EMS Tweezers Style 4, SA steel, Extra fine with superior finish Tips
SDP
Catalog No. 5023861
Encompass_Preferred
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish. Recommended for handling of EM grids.
Material: SA Steel
Tips: Extra fine with superior finish
OAL: 110 mm
Catalog No. 50-238-61 Supplier Electron Microscopy Sciences Supplier No. 783194
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish. Recommended for handling of EM grids.
Material: SA Steel
Tips: Extra fine with superior finish
OAL: 110 mm