Learn More
Electron Microscopy Sciences EMS Tweezers Style 4, NC Ultra-Fine, Extra fine with superior finish Tips
SDP
Catalog No. 5023859
Encompass_Preferred
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish.
Grade: NC Ultra-Fine
Tips: Extra fine with superior finish
OAL: 110 mm
Catalog No. 50-238-59 Supplier Electron Microscopy Sciences Supplier No. 785194
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish.
Grade: NC Ultra-Fine
Tips: Extra fine with superior finish
OAL: 110 mm