Learn More
Electron Microscopy Sciences EMS Wafer Tweezers Style 4WF
SDP
Catalog No. 5023987
Encompass_Preferred
These tweezers are especially designed for the handling of delicate and fragile silicon wafers, scintillation discs, glass slides, coverslips etc. Made from antimagnetic/anti acid steel with a very smooth surface and a non-glare finish.
Tip Width: 12.0 mm
Depth: 9.0 mm
Tip Lip: E:2.5 mm
Offset: 4.0 mm
Catalog No. 50-239-87 Supplier Electron Microscopy Sciences Supplier No. 784144WF
These tweezers are especially designed for the handling of delicate and fragile silicon wafers, scintillation discs, glass slides, coverslips etc. Made from antimagnetic/anti acid steel with a very smooth surface and a non-glare finish.
Tip Width: 12.0 mm
Depth: 9.0 mm
Tip Lip: E:2.5 mm
Offset: 4.0 mm