Learn More
Electron Microscopy Sciences EMS Wafer Tweezers Style 4WFC Pr
SDP
Catalog No. 5023995
Encompass_Preferred
These tweezers are especially designed for the handling of delicate and fragile silicon wafers, scintillation discs, glass slides, coverslips etc. Made from antimagnetic/anti acid steel with a very smooth surface and a non-glare finish. Tips are PEEK/CF30. Ideal for gently holding wafers.
Tip Width: 12 mm
Depth: 2.0 mm
Catalog No. 50-239-95 Supplier Electron Microscopy Sciences Supplier No. 784144WFC
These tweezers are especially designed for the handling of delicate and fragile silicon wafers, scintillation discs, glass slides, coverslips etc. Made from antimagnetic/anti acid steel with a very smooth surface and a non-glare finish. Tips are PEEK/CF30. Ideal for gently holding wafers.
Tip Width: 12 mm
Depth: 2.0 mm