Learn More
Electron Microscopy Sciences EMS Tweezers Style 5, Gold Plating, Extra fine with superior finish Tips
SDP
Catalog No. 5023901
Encompass_Preferred
The tweezers are the Swiss highest quality.
Material: Gold Plating
Tips: Extra fine with superior finish
OAL: 110 mm
Catalog No. 50-239-01 Supplier Electron Microscopy Sciences Supplier No. 783205GP
The tweezers are the Swiss highest quality.
Material: Gold Plating
Tips: Extra fine with superior finish
OAL: 110 mm