Learn More
Electron Microscopy Sciences EMS Tweezers Style 5, Titanium alloy, Extra fine with superior finish Tips
SDP
Catalog No. 5023904
Encompass_Preferred
The tweezers are the Swiss highest quality. Titanium alloy has a relatively low density, good mechanical properties and high use temperature (1600°F/870°C). It is non magnetic, has good corrosion resistance, and a very high strength-to weight ratio.
Material: Titanium Alloy
Tips: Extra fine with superior finish
OAL: 110 mm
Catalog No. 50-239-04 Supplier Electron Microscopy Sciences Supplier No. 783205TI
The tweezers are the Swiss highest quality. Titanium alloy has a relatively low density, good mechanical properties and high use temperature (1600°F/870°C). It is non magnetic, has good corrosion resistance, and a very high strength-to weight ratio.
Material: Titanium Alloy
Tips: Extra fine with superior finish
OAL: 110 mm