Learn More
Electron Microscopy Sciences EMS Tweezers Style 51S, SA steel, Extra fine, bent tips Tips
SDP
Catalog No. 5023889
Encompass_Preferred
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish. Useful for handling EM grids.
Material: SA Steel
Tips: Extra fine, bent tips
OAL: 115 mm
Catalog No. 50-238-89 Supplier Electron Microscopy Sciences Supplier No. 7834051S
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish. Useful for handling EM grids.
Material: SA Steel
Tips: Extra fine, bent tips
OAL: 115 mm