Learn More
Electron Microscopy Sciences EMS Flat-Tip Tweezers Style 574A, 145 mm OAL
SDP
Supplier:  Electron Microscopy Sciences 7821201
Encompass_Preferred
Flat-Tip Tweezers are made from SA low carbon austenitic steel that is non magnetic and anti acid which is ideal for use where corrosion resistance and toughness are primary requirements. The tweezers have serrated grips. Flat and tips bent upward. Tip width: 6mm.
OAL: 145 mm
Catalog No. 50-239-64