Learn More
Electron Microscopy Sciences EMS Tweezers Style 5B, SA steel, Extra fine, bent tips, maxium visibility Tips
SDP
Catalog No. 5023899
Encompass_Preferred
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish.
Material: SA Steel
Tips: Extra fine, bent tips, maxium visibility
OAL: 110 mm
Catalog No. 50-238-99 Supplier Electron Microscopy Sciences Supplier No. 783205B
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish.
Material: SA Steel
Tips: Extra fine, bent tips, maxium visibility
OAL: 110 mm