Learn More
Electron Microscopy Sciences EMS Tweezers Style 5C, SA steel, Extra fine, double bent tips Tips
SDP
Catalog No. 5023900
Encompass_Preferred
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish.
Material: SA Steel
Tips: Extra fine, double bent tips
OAL: 115 mm
Catalog No. 50-239-00 Supplier Electron Microscopy Sciences Supplier No. 783205C
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish.
Material: SA Steel
Tips: Extra fine, double bent tips
OAL: 115 mm