Learn More
Electron Microscopy Sciences EMS Tweezers Style 5TTH, SA steel, Anti-capillary, extra fine tips with locking ring Tips
SDP
Catalog No. 5023897
Encompass_Preferred
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish. Useful for handling EM grids.
Material: SA Steel
Tips: Anti-capillary, extra fine tips with locking ring
OAL: 110 mm
Catalog No. 50-238-97 Supplier Electron Microscopy Sciences Supplier No. 783205TTH
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish. Useful for handling EM grids.
Material: SA Steel
Tips: Anti-capillary, extra fine tips with locking ring
OAL: 110 mm