Learn More
Electron Microscopy Sciences EMS Tweezers Style 5X, SA steel, Extra fine tips, self closing Tips
SDP
Catalog No. 5023905
Encompass_Preferred
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish.
Material: SA Steel
Tips: Extra fine tips, self closing
OAL: 110 mm
Catalog No. 50-239-05 Supplier Electron Microscopy Sciences Supplier No. 783205X
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish.
Material: SA Steel
Tips: Extra fine tips, self closing
OAL: 110 mm