Learn More
Electron Microscopy Sciences EMS Tweezers Style 6, SA steel, Fine, angled tips Tips
SDP
Catalog No. 5023907
Encompass_Preferred
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish.
Material: SA Steel
Tips: Fine, angled tips
OAL: 115 mm
Catalog No. 50-239-07 Supplier Electron Microscopy Sciences Supplier No. 783216
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish.
Material: SA Steel
Tips: Fine, angled tips
OAL: 115 mm