Learn More
Electron Microscopy Sciences EMS Tweezers Style 62A, SA steel, Flat, blunt tips Tips
SDP
Catalog No. 5023892
Encompass_Preferred
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish.
Material: SA Steel
Tips: Flat, blunt tips
OAL: 120 mm
Catalog No. 50-238-92 Supplier Electron Microscopy Sciences Supplier No. 7835062A
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish.
Material: SA Steel
Tips: Flat, blunt tips
OAL: 120 mm