Learn More
Electron Microscopy Sciences EMS Super Thin and Long Tweezers Style 64A, 140 mm OAL
SDP
Catalog No. 5023951
Encompass_Preferred
These long and slender tweezers work well in multiple precision applications and are especially well suited for working near heat sources. They are made from SA low carbon austenitic steel that is non magnetic and anti acid and is ideal for use where corrosion resistance and toughness are primary requirements. Sharp, curve tips.
OAL: 140 mm
Catalog No. 50-239-51 Supplier Electron Microscopy Sciences Supplier No. 7835164A
These long and slender tweezers work well in multiple precision applications and are especially well suited for working near heat sources. They are made from SA low carbon austenitic steel that is non magnetic and anti acid and is ideal for use where corrosion resistance and toughness are primary requirements. Sharp, curve tips.
OAL: 140 mm