Learn More
Electron Microscopy Sciences EMS Tweezers Style 7, SA steel, Fine Curved tips Tips
SDP
Catalog No. NC9917040
Encompass_Preferred
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish.
Material: SA Steel
Tips: Fine Curved tips
OAL: 115 mm
Catalog No. NC9917040 Supplier Electron Microscopy Sciences Supplier No. 783227
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish.
Material: SA Steel
Tips: Fine Curved tips
OAL: 115 mm