Learn More
Electron Microscopy Sciences EMS Tweezers Style 7, PTFE Coating, Fine Curved tips Tips
SDP
Catalog No. 5023886
Encompass_Preferred
The tweezers are the Swiss highest quality. PTFE is a superior anti-scratch, non-stick material.
Material: PTFE Coating
Tips: Fine Curved tips
OAL: 115 mm
Catalog No. 50-238-86 Supplier Electron Microscopy Sciences Supplier No. 783227TE
The tweezers are the Swiss highest quality. PTFE is a superior anti-scratch, non-stick material.
Material: PTFE Coating
Tips: Fine Curved tips
OAL: 115 mm