Learn More
Electron Microscopy Sciences EMS Tweezers Style 7B, SA steel, Fine, curved and serrated tips Tips
SDP
Catalog No. 5023896
Encompass_Preferred
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish.
Material: SA Steel
Tips: Fine, curved and serrated tips
OAL: 115 mm
Catalog No. 50-238-96 Supplier Electron Microscopy Sciences Supplier No. 783227B
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish.
Material: SA Steel
Tips: Fine, curved and serrated tips
OAL: 115 mm