Learn More
Electron Microscopy Sciences EMS Wafer Tweezers Style 8WF
SDP
Catalog No. 5024003
Encompass_Preferred
These tweezers are especially designed for the handling of delicate and fragile silicon wafers, scintillation discs, glass slides, coverslips etc. Made from antimagnetic/anti acid steel with a very smooth surface and a non-glare finish.
Tip Width: 28.5 mm
Depth: 12.0 mm
Tip Lip: 2.2 mm
Offset: 3.5 mm
Catalog No. 50-240-03 Supplier Electron Microscopy Sciences Supplier No. 784208WF
These tweezers are especially designed for the handling of delicate and fragile silicon wafers, scintillation discs, glass slides, coverslips etc. Made from antimagnetic/anti acid steel with a very smooth surface and a non-glare finish.
Tip Width: 28.5 mm
Depth: 12.0 mm
Tip Lip: 2.2 mm
Offset: 3.5 mm