Learn More
Electron Microscopy Sciences EMS Wafer Tweezers Style 8WNY
SDP
Catalog No. 5023992
Encompass_Preferred
These tweezers are especially designed for the handling of delicate and fragile silicon wafers, scintillation discs, glass slides, coverslips etc. Made from antimagnetic/anti acid steel with a very smooth surface and a non-glare finish.
Tip Width: 65.0 mm
Depth: 19.0 mm
Tip Lip: 1.3 mm
Offset: 2.0 mm
Catalog No. 50-239-92 Supplier Electron Microscopy Sciences Supplier No. 784228WNY
These tweezers are especially designed for the handling of delicate and fragile silicon wafers, scintillation discs, glass slides, coverslips etc. Made from antimagnetic/anti acid steel with a very smooth surface and a non-glare finish.
Tip Width: 65.0 mm
Depth: 19.0 mm
Tip Lip: 1.3 mm
Offset: 2.0 mm