Learn More
Electron Microscopy Sciences EMS Tweezers Style OA, SA steel, Straight point with radiused outside edges, fine tips Tips
SDP
Catalog No. 5023882
Encompass_Preferred
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel is used where corrosion resistance and toughness are primary requirements. Anti-glare satin finish.
Material: SA Steel
Tips: Straight point with radiused outside edges, fine tips
OAL: 120 mm
Catalog No. 50-238-82 Supplier Electron Microscopy Sciences Supplier No. 72919OA
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel is used where corrosion resistance and toughness are primary requirements. Anti-glare satin finish.
Material: SA Steel
Tips: Straight point with radiused outside edges, fine tips
OAL: 120 mm