Learn More
Electron Microscopy Sciences EMS Super Thin and Long Tweezers Style SSL, 136 mm OAL
SDP
Catalog No. 5023947
Encompass_Preferred
These long and slender tweezers work well in multiple precision applications and are especially well suited for working near heat sources. They are made from SA low carbon austenitic steel that is non magnetic and anti acid and is ideal for use where corrosion resistance and toughness are primary requirements. Long and narrow tips, ergonomic shape.
OAL: 136 mm
Catalog No. 50-239-47 Supplier Electron Microscopy Sciences Supplier No. 78353SSL
These long and slender tweezers work well in multiple precision applications and are especially well suited for working near heat sources. They are made from SA low carbon austenitic steel that is non magnetic and anti acid and is ideal for use where corrosion resistance and toughness are primary requirements. Long and narrow tips, ergonomic shape.
OAL: 136 mm