Learn More
Electron Microscopy Sciences Ultramicrotome Aid
SDP
Catalog No. 5034061
Encompass_Preferred
Made from hard plastic (Delrin) with a beautifully machined finish to handle many of your instruments that need to be at your fingertips during sectioning. This EMS Ultra-microtome Aid is a safe place to rest your diamond knife, diamond  trimming tool, tweezers, hair curl or eye lash, diamond knife cleaning rods, perfect-loop for picking up sections. Note: This unit is not a safe device to store your sectioning knives, such as diamond or glass knives, after you're done the sectioning.
Catalog No. 50-340-61 Supplier Electron Microscopy Sciences Supplier No. 70612
Made from hard plastic (Delrin) with a beautifully machined finish to handle many of your instruments that need to be at your fingertips during sectioning. This EMS Ultra-microtome Aid is a safe place to rest your diamond knife, diamond  trimming tool, tweezers, hair curl or eye lash, diamond knife cleaning rods, perfect-loop for picking up sections. Note: This unit is not a safe device to store your sectioning knives, such as diamond or glass knives, after you're done the sectioning.