Learn More
Medchemexpress LLC EMT inhibitor-2 | 2232228-60-1 | 98.8% | 25 MG
SDP
Catalog No. 5000341391
Encompass_Preferred
EMT inhibitor-2 (Compound 1) inhibits epithelial-mesenchymal transition (EMT) induced by substances such as IL-1β and TGF-β released from immunocytes. It also inhibits CYP3A4 testosterone and CYP2C9 with IC50s of 49.72 and 5.54 μM, respectively.
  • Inhibits epithelial-mesenchymal transition (EMT)
  • Inhibits CYP3A4 testosterone
  • Inhibits CYP2C9
Catalog No. 50-003-41391 Supplier Medchemexpress LLC Supplier No. 25MGHY128859
EMT inhibitor-2 (Compound 1) inhibits epithelial-mesenchymal transition (EMT) induced by substances such as IL-1β and TGF-β released from immunocytes. It also inhibits CYP3A4 testosterone and CYP2C9 with IC50s of 49.72 and 5.54 μM, respectively.
  • Inhibits epithelial-mesenchymal transition (EMT)
  • Inhibits CYP3A4 testosterone
  • Inhibits CYP2C9