Learn More
Medchemexpress LLC Emtricitabine-13C,15N2 | 1217820-69-3 | 250.23 | 500 UG
SDP
Catalog No. 5000386246
Encompass_Preferred
Emtricitabine-13C,15N2 is a 13C- and 15N-labeled Emtricitabine. It is a nucleoside reverse transcriptase inhibitor (NRTI) with an EC50 of 0.01 μM in PBMC cell and acts as an antiviral agent for the treatment of HIV infection. This product is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-86246 Supplier Medchemexpress LLC Supplier No. HY17427S2500UG
Emtricitabine-13C,15N2 is a 13C- and 15N-labeled Emtricitabine. It is a nucleoside reverse transcriptase inhibitor (NRTI) with an EC50 of 0.01 μM in PBMC cell and acts as an antiviral agent for the treatment of HIV infection. This product is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS