Learn More
Medchemexpress LLC Emtricitabine-15N,d2 | 2714436-53-8 | 97.6% | 250.25 | 1 MG
SDP
Catalog No. 5000386619
Encompass_Preferred
Emtricitabine-15N,d2 is a 15N-labeled and deuterium labeled Emtricitabine. Emtricitabine is a nucleoside reverse transcriptase inhibitor (NRTI) with an EC50 of 0.01 μM in PBMC cell. It is an antiviral agent for the treatment of HIV infection.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-86619 Supplier Medchemexpress LLC Supplier No. HY17427S1MG
Emtricitabine-15N,d2 is a 15N-labeled and deuterium labeled Emtricitabine. Emtricitabine is a nucleoside reverse transcriptase inhibitor (NRTI) with an EC50 of 0.01 μM in PBMC cell. It is an antiviral agent for the treatment of HIV infection.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.