Learn More
Medchemexpress LLC HY-17427 500mg , Emtricitabine BW1592 CAS:143491-57-0 Purity:98%
SDP
Catalog No. 501492510
Encompass_Preferred
Medchemexpress, HY-17427 500mg Emtricitabine BW1592 CAS:143491-57-0 Formula:C8H10FN3O3S EC50: 0.01 µM (NRTI, PBMC), 0.026 µM (NRTI, HeLa cell) Purity:98% Emtricitabine is an inhibitor of the nucleoside reverse transcriptase ( NRTI ) and human immunodeficiency virus type 1 ( HIV-1 ); inhibits NRTI with an EC 50 of 0.01 µM in PBMC cell. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2510 Supplier Medchemexpress LLC Supplier No. HY17427500MG
Medchemexpress, HY-17427 500mg Emtricitabine BW1592 CAS:143491-57-0 Formula:C8H10FN3O3S EC50: 0.01 µM (NRTI, PBMC), 0.026 µM (NRTI, HeLa cell) Purity:98% Emtricitabine is an inhibitor of the nucleoside reverse transcriptase ( NRTI ) and human immunodeficiency virus type 1 ( HIV-1 ); inhibits NRTI with an EC 50 of 0.01 µM in PBMC cell. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.