Learn More
Selleck Chemical LLC Emtricitabine 50mg 143491-57-0
SDP
Catalog No. 505900
Encompass_Preferred
Emtricitabine (FTC) is a new nucleoside agent that has activity against both human immunodeficiency virus (HIV) and hepatitis B virus. It is a reverse transcriptase inhibitor. Intracellular half-life is 39 h. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-590-0 Supplier Selleck Chemical LLC Supplier No. S170450MG
Emtricitabine (FTC) is a new nucleoside agent that has activity against both human immunodeficiency virus (HIV) and hepatitis B virus. It is a reverse transcriptase inhibitor. Intracellular half-life is 39 h. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.