Learn More
Medchemexpress LLC EMU-116 | 2241724-59-2 | 98.5% | 405.58 | 50 MG
SDP
Catalog No. 5000390512
Encompass_Preferred
EMU-116 is an orally active CXCR4 antagonist that can be used in the study of cancer. It is provided in various pack sizes, including 50 mg. This compound is intended for research use only.
  • Orally active CXCR4 antagonist
  • Used in the study of cancer
  • High purity compound
Catalog No. 50-003-90512 Supplier Medchemexpress LLC Supplier No. HY16853150MG
EMU-116 is an orally active CXCR4 antagonist that can be used in the study of cancer. It is provided in various pack sizes, including 50 mg. This compound is intended for research use only.
  • Orally active CXCR4 antagonist
  • Used in the study of cancer
  • High purity compound