Learn More
MSC Emuge Plug Thread Gage, 3/4-10 Thread, 2B Class, Double End, Go/No Go
Catalog No. 5000533937
Encompass
This plug thread gage is designed for precise thread inspection. It features a double-ended design, providing both Go and No Go functionality for 3/4-10 threads with a 2B class. Constructed from durable tool steel, it meets ASME B1.2 standards, ensuring reliable and accurate measurements for quality control.
  • Go/No Go functionality
  • 3/4-10 Thread size
  • Tool steel material
  • 2B Classification
  • Handle included
  • Conforms to ASME B1.2 standards
  • Right hand threading
  • Bright/uncoated finish
  • Double end design
  • Includes short form certification
  • Country of origin: Germany
Catalog No. 50-005-33937 Supplier MSC Supplier No. 16925737
This plug thread gage is designed for precise thread inspection. It features a double-ended design, providing both Go and No Go functionality for 3/4-10 threads with a 2B class. Constructed from durable tool steel, it meets ASME B1.2 standards, ensuring reliable and accurate measurements for quality control.
  • Go/No Go functionality
  • 3/4-10 Thread size
  • Tool steel material
  • 2B Classification
  • Handle included
  • Conforms to ASME B1.2 standards
  • Right hand threading
  • Bright/uncoated finish
  • Double end design
  • Includes short form certification
  • Country of origin: Germany