Learn More
MSC Emuge Plug Thread Gage, M12x1 Thread, 6H Class, Double End, Go/No-Go
Catalog No. 5000532642
Encompass
This plug thread gage is designed for M12x1 threads with a 6H class. It features a double end with both Go and No Go functionalities and includes a handle. The gage is constructed from tool steel, has a bright/uncoated finish, and adheres to DIN ISO 1502 standards, ensuring precise thread measurement. It also comes with a short form certification and is suitable for right-hand threading.
  • Go and no go functionality
  • Designed for M12x1 threads
  • Made from tool steel
  • 6H class classification
  • Handle included
  • Adheres to DIN ISO 1502 standards
  • Suitable for right hand threading
  • Bright/uncoated finish
  • Double end design
  • Includes short form certification
  • Manufactured in Germany
Catalog No. 50-005-32642 Supplier MSC Supplier No. 16924821
This plug thread gage is designed for M12x1 threads with a 6H class. It features a double end with both Go and No Go functionalities and includes a handle. The gage is constructed from tool steel, has a bright/uncoated finish, and adheres to DIN ISO 1502 standards, ensuring precise thread measurement. It also comes with a short form certification and is suitable for right-hand threading.
  • Go and no go functionality
  • Designed for M12x1 threads
  • Made from tool steel
  • 6H class classification
  • Handle included
  • Adheres to DIN ISO 1502 standards
  • Suitable for right hand threading
  • Bright/uncoated finish
  • Double end design
  • Includes short form certification
  • Manufactured in Germany