Learn More
MSC Emuge Plug Thread Gage, M20x1 Thread, 6H Class, Double End, Go/No Go
Catalog No. 5000529220
Encompass
This plug thread gage features an M20x1 thread with a 6H class. It is a double-ended design, incorporating both "Go" and "No Go" sides for precise measurement, and includes a handle for ease of use. Manufactured from durable tool steel with a bright/uncoated finish, it is designed for right-hand threads and comes with short form certification. This product is manufactured in Germany.
  • Go and No Go measurement
  • M20x1 thread size
  • Tool steel material
  • 6H classification
  • Handle included
  • Conforms to DIN ISO 1502 standards
  • Right hand threading
  • Bright/uncoated finish
  • Double end design
  • Includes short form certification
  • Country of origin: Germany
Catalog No. 50-005-29220 Supplier MSC Supplier No. 16925273
This plug thread gage features an M20x1 thread with a 6H class. It is a double-ended design, incorporating both "Go" and "No Go" sides for precise measurement, and includes a handle for ease of use. Manufactured from durable tool steel with a bright/uncoated finish, it is designed for right-hand threads and comes with short form certification. This product is manufactured in Germany.
  • Go and No Go measurement
  • M20x1 thread size
  • Tool steel material
  • 6H classification
  • Handle included
  • Conforms to DIN ISO 1502 standards
  • Right hand threading
  • Bright/uncoated finish
  • Double end design
  • Includes short form certification
  • Country of origin: Germany