Learn More
MSC Emuge Plug Thread Gage, M3x0.5 Thread, 6H Class, Double End, Tool Steel
Catalog No. 5000529354
Encompass
This plug thread gage features an M3x0.5 thread and 6H class. It is a double-ended gage with both "Go" and "No Go" ends, and a handle is included. Constructed from tool steel, it conforms to DIN ISO 1502 standards. The gage has right-hand threading and a bright/uncoated finish, and includes short form certification.
  • Double end, go & no go
  • Handle included
  • Made of tool steel
  • Conforms to DIN ISO 1502 standards
  • Includes short form certification
  • Country of origin: Germany
Catalog No. 50-005-29354 Supplier MSC Supplier No. 16924961
This plug thread gage features an M3x0.5 thread and 6H class. It is a double-ended gage with both "Go" and "No Go" ends, and a handle is included. Constructed from tool steel, it conforms to DIN ISO 1502 standards. The gage has right-hand threading and a bright/uncoated finish, and includes short form certification.
  • Double end, go & no go
  • Handle included
  • Made of tool steel
  • Conforms to DIN ISO 1502 standards
  • Includes short form certification
  • Country of origin: Germany