Learn More
MSC Emuge Plug Thread Gage, M4x0.7, 6H, Double End, Go/No Go
Catalog No. 5000513133
Encompass
This plug thread gage is a double-ended tool designed for precise inspection of M4x0.7 threads. It features both a "Go" and a "No Go" end, ensuring accurate verification of internal thread limits according to DIN ISO 1502 standards with a 6H class. Constructed from durable tool steel, it provides reliable performance and comes with a handle for convenient use.
  • Double-ended design with both "Go" and "No Go" functions.
  • Designed for M4x0.7 threads with 6H class.
  • Manufactured from durable tool steel.
  • Conforms to DIN ISO 1502 standards.
  • Includes a handle for ease of use.
  • Bright/uncoated finish.
  • Right-hand threading.
  • Includes short form certification.
  • Country of origin: Germany.
Catalog No. 50-005-13133 Supplier MSC Supplier No. 16925000
This plug thread gage is a double-ended tool designed for precise inspection of M4x0.7 threads. It features both a "Go" and a "No Go" end, ensuring accurate verification of internal thread limits according to DIN ISO 1502 standards with a 6H class. Constructed from durable tool steel, it provides reliable performance and comes with a handle for convenient use.
  • Double-ended design with both "Go" and "No Go" functions.
  • Designed for M4x0.7 threads with 6H class.
  • Manufactured from durable tool steel.
  • Conforms to DIN ISO 1502 standards.
  • Includes a handle for ease of use.
  • Bright/uncoated finish.
  • Right-hand threading.
  • Includes short form certification.
  • Country of origin: Germany.