Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Emulsifying Wax, NF, Spectrum™ Chemical
SDP

Catalog No. 18602408 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
12 x 500 g
25 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-408 25 kg Poly Bottle
18-602-406 500 g Poly Bottle
18-602-407 12 x 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-602-408 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. W102625KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

W1026

Spectrum™ Chemical Emulsifying Wax, NF is used as a thickening agent excipent. All Spectrum Chemical NF products are packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

CAS 8005-44-5
CAS Max % 100%
Melting Range 50°C to 54°C
Packaging Poly Bottle
Quantity 25 kg
Grade NF
Physical Form Powder or Waxy Solid
Chemical Name or Material Emulsifying Wax
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.