Learn More
Perkin Elmer US LLC Antifoaming Silicone Emulsion, Dow Corning DB 110 A, 500mL
Catalog No. 509042471
Encompass_Preferred
Controls foaming during reaction when using the MHS-10 or MHS-20.
Catalog No. 50-904-2471 Supplier Perkin Elmer US LLC Supplier No. B0507226
Controls foaming during reaction when using the MHS-10 or MHS-20.