Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Emvododstat (PTC299) | 1256565-36-2 | 99.9% | 467.34 g/mol | C25H20Cl2N2O3 | 50MG
SDP

Catalog No. 5000237023
Encompass_Preferred

PTC299 (emvododstat) is an orally active small-molecule inhibitor that selectively suppresses VEGFA mRNA translation and inhibits dihydroorotate dehydrogenase (DHODH). Provided for research use, it is supplied as solid samples and DMSO solutions and is used in studies of VEGF regulation, pyrimidine biosynthesis, and oncology and immunology research.

  • High purity: 99.9% by assay.
  • Molecular weight: 467.34 g/mol.
  • Chemical formula: C25H20Cl2N2O3.
  • Physical appearance: white to light yellow solid.
  • Available as solid and as 10 mM solution in DMSO.
  • Suitable for studies of VEGF regulation and DHODH-related pathways.
  • Offered in multiple package sizes including 50 mg.

Catalog No. 50-002-37023 Supplier Medchemexpress LLC Supplier No. HY12459350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PTC299 (emvododstat) is an orally active small-molecule inhibitor that selectively suppresses VEGFA mRNA translation and inhibits dihydroorotate dehydrogenase (DHODH). Provided for research use, it is supplied as solid samples and DMSO solutions and is used in studies of VEGF regulation, pyrimidine biosynthesis, and oncology and immunology research.

  • High purity: 99.9% by assay.
  • Molecular weight: 467.34 g/mol.
  • Chemical formula: C25H20Cl2N2O3.
  • Physical appearance: white to light yellow solid.
  • Available as solid and as 10 mM solution in DMSO.
  • Suitable for studies of VEGF regulation and DHODH-related pathways.
  • Offered in multiple package sizes including 50 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.