Learn More
Aviva Systems Biology Corp Emx1 Antibody - middle region (ARP51624_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51624_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Emx1. It was validated on Western Blot. . Alternate names: MGC129284, MGC129285ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-580