Learn More
Aviva Systems Biology Corp EMX2 Antibody - N-terminal region (ARP31643_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31643_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EMX2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Human EMX2. Alternate names: -ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54293