Learn More
Meiji Techno America Meiji Techno America Binocular Zoom Stereo microscope body with zoom range, W.D. 93mm magnification from 7X to 90X (Using SWF10X and SWF20X eyepieces)
Catalog No. NC0836907
Encompass_Preferred
Binocular Zoom Stereo microscope body with zoom range, W.D. 93mm magnification from 7X to 90X (Using SWF10X and SWF20X eyepieces)
Catalog No. NC0836907 Supplier Meiji Techno America Supplier No. EMZ5
Binocular Zoom Stereo microscope body with zoom range, W.D. 93mm magnification from 7X to 90X (Using SWF10X and SWF20X eyepieces)