Learn More
Aviva Systems Biology Corp EN2 Antibody - C-terminal region (ARP31375_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31375_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EN2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human EN2. Alternate names: -ApplicationIHC, WB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-54199