Learn More
TARGETMOL CHEMICALS INC EN4 25MG
SDP
Catalog No. 502579024
Encompass_Preferred
Also available in 1 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. EN4 (EN4 MYC inhibitor) MYC inhibitor is a covalent ligand that targets cysteine 171 (C171) of MYC. It is selective for c-MYC over N-MYC and L-MYC and inhibits MYC transcriptional activity downregulates MYC targets and impairs tumorigenesis. purity: 98%
Catalog No. 50-257-9024 Supplier TARGETMOL CHEMICALS INC Supplier No. T906125MG
Also available in 1 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. EN4 (EN4 MYC inhibitor) MYC inhibitor is a covalent ligand that targets cysteine 171 (C171) of MYC. It is selective for c-MYC over N-MYC and L-MYC and inhibits MYC transcriptional activity downregulates MYC targets and impairs tumorigenesis. purity: 98%