Learn More
Selleck Chemical LLC EN4 5mg
SDP
Supplier:  Selleck Chemical LLC S98075MG
Encompass_Preferred
EN4, a covalent ligand that targets cysteine 171 (C171) of MYC, is selective for c-MYC over N-MYC and L-MYC. EN4 inhibits MYC transcriptional activity, downregulates MYC targets, and impairs tumorigenesis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3510