Learn More
TARGETMOL CHEMICALS INC EN523 5MG
SDP
Catalog No. 502578941
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. EN523 targets non-catalytic allosteric cysteine C23 in K48 ubiquitin-specific deuquitinase OTUB1. purity: 98%
Catalog No. 50-257-8941 Supplier TARGETMOL CHEMICALS INC Supplier No. T98845MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. EN523 targets non-catalytic allosteric cysteine C23 in K48 ubiquitin-specific deuquitinase OTUB1. purity: 98%